Assay Details
Gene Symbol: Slc26a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177243.4 | NP_796217.2 | 19 - 20 | 2369 | 74 | |
GenBank mRNA | AK078966.1 | - | 19 - 20 | 2370 | 74 | |
AK086815.1 | - | 19 - 20 | 2370 | 74 | ||
AY034145.1 | - | 18 - 19 | 2282 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
