Assay Details
Gene Symbol: Il6st | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010560.3 | NP_034690.3 | 7 - 8 | 1087 | 109 | |
GenBank mRNA | BC020454.1 | - | 7 - 8 | 1041 | 109 | |
BC058679.1 | - | 7 - 8 | 1048 | 6834623 | 109 | |
M83336.1 | - | 6 - 7 | 892 | 109 | ||
X62646.1 | - | 6 - 7 | 892 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
