Assay Details
Gene Symbol: Gphn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145965.2 | NP_666077.2 | 3 - 4 | 326 | 119 | |
NM_172952.3 | NP_766540.2 | 3 - 4 | 326 | 119 | ||
GenBank mRNA | AK082353.1 | - | 3 - 4 | 248 | 119 | |
AK144569.1 | - | 3 - 4 | 329 | 119 | ||
BC127058.1 | - | 3 - 4 | 248 | 40109771 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
