Assay Details
Gene Symbol: Lmbrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177178.3 | NP_796152.2 | 3 - 4 | 498 | 63 | |
GenBank mRNA | AK036527.1 | - | 3 - 4 | 519 | 63 | |
AK079450.1 | - | 3 - 4 | 498 | 63 | ||
AK153728.1 | - | 3 - 4 | 449 | 63 | ||
BC095971.1 | - | 3 - 4 | 415 | 30108856 | 63 | |
BC120727.1 | - | 3 - 4 | 470 | 40129650 | 63 | |
BC125421.1 | - | 3 - 4 | 470 | 40129936 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
