Assay Details
Gene Symbol: Brip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178309.2 | NP_840094.1 | 4 - 5 | 493 | 117 | |
GenBank mRNA | AK033205.1 | - | 4 - 5 | 465 | 117 | |
AK051878.1 | - | 4 - 5 | 494 | 117 | ||
AK145709.1 | - | 4 - 5 | 494 | 117 | ||
AK169440.1 | - | 4 - 5 | 440 | 117 | ||
AK169823.1 | - | 4 - 5 | 452 | 117 | ||
BC094252.1 | - | 4 - 5 | 442 | 6401109 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
