Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700012626.mm.1 |
TC1700001403.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012626.mm.1 |
NM_001081416.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012626.mm.1 |
NM_001081416.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012622.mm.1 |
TC1700001403.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012622.mm.1 |
NM_001081416.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012622.mm.1 |
NM_001081416.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012633.mm.1 |
TC1700001403.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012633.mm.1 |
NM_001081416.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012633.mm.1 |
NM_001081416.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012625.mm.1 |
TC1700001403.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012625.mm.1 |
NM_001081416.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012625.mm.1 |
NM_001081416.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012620.mm.1 |
TC1700001403.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012620.mm.1 |
NM_001081416.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012620.mm.1 |
NM_001081416.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012630.mm.1 |
TC1700001403.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012630.mm.1 |
NM_001081416.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012630.mm.1 |
NM_001081416.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012621.mm.1 |
TC1700001403.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012621.mm.1 |
NM_001081416.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012621.mm.1 |
NM_001081416.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012628.mm.1 |
TC1700001403.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012628.mm.1 |
NM_001081416.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012628.mm.1 |
NM_001081416.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012623.mm.1 |
TC1700001403.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012623.mm.1 |
NM_001081416.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012623.mm.1 |
NM_001081416.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012624.mm.1 |
TC1700001403.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012624.mm.1 |
NM_001081416.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012624.mm.1 |
NM_001081416.1 |
Refseq |