Assay Details
Gene Symbol: Atad5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029856.2 | NP_001025027.1 | 10 - 11 | 3464 | 115 | |
GenBank mRNA | AY557610.1 | - | 10 - 11 | 3455 | 115 | |
BC046463.1 | - | 6 - 7 | 761 | 115 | ||
BC141180.1 | - | 9 - 10 | 2960 | 9055792 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
