Assay Details
Gene Symbol: Atad5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029856.2 | NP_001025027.1 | 14 - 15 | 3929 | 126 | |
GenBank mRNA | AY557610.1 | - | 14 - 15 | 3920 | 126 | |
BC046463.1 | - | 10 - 11 | 1226 | 126 | ||
BC141180.1 | - | 13 - 14 | 3425 | 9055792 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
