Assay Details
Gene Symbol: Atad5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029856.2 | NP_001025027.1 | 3 - 4 | 2400 | 92 | |
GenBank mRNA | AK048355.1 | - | 3 - 4 | 2362 | 92 | |
AK085211.1 | - | 3 - 4 | 2391 | 92 | ||
AK165945.1 | - | 3 - 4 | 2367 | 92 | ||
AY557610.1 | - | 3 - 4 | 2391 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
