Assay Details
Gene Symbol: Slc22a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039371.2 | NP_001034460.2 | 11 - 12 | 1889 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473940 | BB017021.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473940 | AK029695.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473940 | NM_001039371.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473940 | BB017021.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473940 | AK029695.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473940 | NM_001039371.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2179738 | BC094925.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2179738 | NM_001039371.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115211 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 522804 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 723852 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 100418 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 433524 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 805580 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 457420 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 928641 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 177452 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115721 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 275920 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781152 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 770511 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 242008 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 631522 | AK045563.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829391 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 807837 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 647719 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 540235 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829328 | AF022072.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829328 | AK012646.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829328 | AK030727.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829328 | AK032927.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829328 | AK163841.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829328 | BC016111.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829328 | BC053842.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829328 | NM_010345.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 191635 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830074 | AK035248.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 611495 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 218926 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 771023 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894017 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 856868 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 415628 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 200994 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 437578 | AK170112.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 437578 | BC027816.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 437578 | BC047218.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 437578 | BC055406.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 406510 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 643559 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 498111 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 644754 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 330184 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 866262 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 544266 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779876 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 401399 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 17308 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 8021 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 92496 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 72230 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 33492 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 102652 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 61426 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110031 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132906 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473940 | BB017021.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473940 | AK029695.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473940 | NM_001039371.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473940 | BB017021.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473940 | AK029695.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473940 | NM_001039371.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2179738 | BC094925.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2179738 | NM_001039371.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132906 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05009921 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05009958 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110031 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05009933 | - | - |
Whole Mouse Genome G4122A | A_52_P483269 | AK029299.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110031 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132906 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05009921 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22378030 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377821 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377868 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22378019 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377770 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03077196 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377826 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377810 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377760 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02565177 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377888 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03023357 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02819688 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377890 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02677542 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377880 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377776 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377876 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03077207 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377757 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02837371 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377829 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377614 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22378027 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22378035 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02380560 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377916 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02666146 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377772 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377805 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22378020 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02296305 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02565171 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377814 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02855903 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377907 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377741 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377744 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02855894 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02876607 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377796 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377798 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02855904 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377884 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377865 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377908 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377617 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02565169 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02989775 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03049632 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377824 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377730 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377733 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377754 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377882 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377842 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377864 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02380563 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377623 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377848 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377859 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02925054 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377736 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377871 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03077205 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377742 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377816 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03191162 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377788 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02876602 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377831 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02666142 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02515090 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02677532 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377873 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02666161 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03191177 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02677527 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22378022 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03191170 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377875 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02888942 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02666144 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02311199 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377735 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377791 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377615 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377722 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02296308 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377613 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22378024 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377780 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377897 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377883 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02434241 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02666138 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03191173 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377782 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03023354 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377619 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03003933 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377745 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377812 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377819 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02855901 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377851 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377867 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02666158 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03077202 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377726 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02888937 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377827 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02311197 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03191175 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02677541 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03077201 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377857 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377904 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377837 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377886 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377818 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02677546 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03077204 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377898 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377799 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02508894 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377616 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377803 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377620 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02611641 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03049633 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02515101 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377909 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377725 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02666163 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22378015 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377815 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02611640 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377917 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03077194 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377763 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22377784 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22378010 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03077190 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10500725 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10500733 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10500722 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10500729 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10500727 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10500731 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10500735 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10500732 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10500726 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10500730 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10500723 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10500728 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10500721 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10500734 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1444312_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1458933_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10500720 | - | - |
Mouse Genome 430 2.0 Array | 1444312_at | - | - |
Mouse Genome 430 2.0 Array | 1458933_at | - | - |
Mouse Genome 430 2.0 Array | 1444312_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1458933_PM_at | - | - |
Mouse Expression Set 430 | 1458933_at | - | - |
Mouse Expression Set 430 | 1444312_PM_at | - | - |
Mouse Expression Set 430 | 1444312_at | - | - |
Mouse Expression Set 430 | 1458933_PM_at | - | - |
Murine Genome U74v2 Set | 131100_at | - | - |
Murine Genome U74v2 Set | 131100_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1444312_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1458933_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1458933_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1444312_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 308907 | - | - |
ABI Mouse Genome Survey Microarray | 581441 | - | - |
