Assay Details
Gene Symbol: Npepps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008942.2 | NP_032968.2 | 2 - 3 | 460 | 152 | |
GenBank mRNA | AK133898.1 | - | 2 - 3 | 268 | 152 | |
AK138772.1 | - | 2 - 3 | 439 | 152 | ||
BC009653.1 | - | 2 - 3 | 399 | 3487224 | 152 | |
BC086798.1 | - | 2 - 3 | 452 | 6812900 | 152 | |
BC098212.1 | - | 2 - 3 | 460 | 6825181 | 152 | |
U35646.1 | - | 2 - 3 | 447 | 152 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
