Assay Details
Gene Symbol: Trpm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134252.3 | NP_599013.1 | 10 - 11 | 1320 | 95 | |
GenBank mRNA | AF481480.2 | - | 1589 | 95 | ||
AY095352.1 | - | 8 - 9 | 1154 | 95 | ||
BC117934.1 | - | 10 - 11 | 1320 | 40097778 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
