Assay Details
Gene Symbol: Trpm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134252.3 | NP_599013.1 | 4 - 5 | 370 | 118 | |
GenBank mRNA | AF481480.2 | - | 639 | 118 | ||
AY095352.1 | - | 2 - 3 | 204 | 118 | ||
BC117934.1 | - | 4 - 5 | 370 | 40097778 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
