Assay Details
Gene Symbol: Zfp598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK138860.1 | - | 3 - 4 | 1126 | 61 | |
BC024690.1 | - | 6 - 7 | 798 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
