Assay Details
Gene Symbol: Lmbr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029098.3 | NP_083374.1 | 2 - 3 | 329 | 75 | |
GenBank mRNA | AK003656.1 | - | 2 - 3 | 261 | 75 | |
AK030589.1 | - | 2 - 3 | 295 | 75 | ||
AK049110.1 | - | 2 - 3 | 330 | 75 | ||
AK143891.1 | - | 2 - 3 | 290 | 75 | ||
AK154968.1 | - | 2 - 3 | 463 | 75 | ||
AK170328.1 | - | 2 - 3 | 290 | 75 | ||
AK173121.1 | - | 2 - 3 | 710 | 75 | ||
AY052398.1 | - | 2 - 3 | 172 | 75 | ||
BC023397.1 | - | 2 - 3 | 266 | 5043877 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
