Assay Details
Gene Symbol: Lmbr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029098.3 | NP_083374.1 | 7 - 8 | 809 | 82 | |
GenBank mRNA | AK003656.1 | - | 7 - 8 | 741 | 82 | |
AK030589.1 | - | 7 - 8 | 775 | 82 | ||
AK049110.1 | - | 7 - 8 | 810 | 82 | ||
AK143891.1 | - | 7 - 8 | 770 | 82 | ||
AK154968.1 | - | 7 - 8 | 943 | 82 | ||
AK170328.1 | - | 7 - 8 | 770 | 82 | ||
AK173121.1 | - | 7 - 8 | 1190 | 82 | ||
AY052398.1 | - | 7 - 8 | 652 | 82 | ||
BC023397.1 | - | 7 - 8 | 746 | 5043877 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
