Assay Details
Gene Symbol: Chrnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021600.3 | NP_067611.2 | 3 - 4 | 343 | 68 | |
GenBank mRNA | BC052153.1 | - | 3 - 4 | 288 | 6336578 | 68 |
BC141376.1 | - | 3 - 4 | 271 | 9055989 | 68 | |
BC141387.1 | - | 3 - 4 | 271 | 9056000 | 68 | |
K02582.1 | - | 3 - 4 | 254 | 68 | ||
L10076.1 | - | 3 - 4 | 303 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
