Assay Details
Gene Symbol: Chrnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021600.3 | NP_067611.2 | 9 - 10 | 1157 | 65 | |
GenBank mRNA | AK081344.1 | - | 8 - 9 | 1143 | 65 | |
BC052153.1 | - | 9 - 10 | 1102 | 6336578 | 65 | |
BC141376.1 | - | 9 - 10 | 1085 | 9055989 | 65 | |
BC141387.1 | - | 9 - 10 | 1085 | 9056000 | 65 | |
K02582.1 | - | 9 - 10 | 1068 | 65 | ||
L10076.1 | - | 9 - 10 | 1117 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
