Assay Details
Gene Symbol: Dennd2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177857.1 | NP_808525.2 | 15 - 16 | 2582 | 80 | |
GenBank mRNA | AK137637.1 | - | 10 - 11 | 1189 | 80 | |
AK172232.1 | - | 11 - 12 | 1304 | 80 | ||
BC046440.1 | - | 11 - 12 | 1050 | 80 | ||
BC060666.1 | - | 15 - 16 | 2582 | 6841982 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
