Assay Details
Gene Symbol: Gas2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013759.2 | NP_001013781.1 | 2 - 3 | 724 | 66 | |
GenBank mRNA | BC147480.1 | - | 2 - 3 | 724 | 9087436 | 66 |
BC150847.1 | - | 2 - 3 | 724 | 9087758 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
