Assay Details
Gene Symbol: Bcl2l15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142959.1 | NP_001136431.1 | 1 - 2 | 277 | 67 | |
NM_001142960.1 | NP_001136432.1 | 1 - 2 | 277 | 67 | ||
GenBank mRNA | AF014453.1 | - | 1 - 2 | 373 | 67 | |
BC027668.1 | - | 1 - 2 | 113 | 67 | ||
BC052523.1 | - | 1 - 2 | 211 | 67 | ||
BC117780.1 | - | 1 - 2 | 277 | 40091931 | 67 | |
BC117781.1 | - | 1 - 2 | 277 | 40091934 | 67 | |
BC128505.1 | - | 1 - 2 | 277 | 40091935 | 67 | |
BC128506.1 | - | 1 - 2 | 277 | 40091936 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
