Assay Details
Gene Symbol: Farp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145519.2 | NP_663494.2 | 3 - 4 | 399 | 85 | |
GenBank mRNA | AK050860.1 | - | 3 - 4 | 359 | 85 | |
AK086699.1 | - | 3 - 4 | 392 | 85 | ||
AK171713.1 | - | 3 - 4 | 425 | 85 | ||
BB663505.1 | - | 3 - 4 | 392 | 85 | ||
BC009153.1 | - | 3 - 4 | 388 | 2655209 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
