Assay Details
Gene Symbol: Tom1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028011.2 | NP_082287.1 | 7 - 8 | 769 | 62 | |
GenBank mRNA | AF395837.1 | - | 7 - 8 | 733 | 62 | |
AJ505007.1 | - | 7 - 8 | 730 | 62 | ||
AK009826.1 | - | 5 - 6 | 504 | 62 | ||
AK033712.1 | - | 7 - 8 | 771 | 62 | ||
AK083630.1 | - | 731 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
