Assay Details
Gene Symbol: A230001M10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK038386.1 | - | 863 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
vang-like 1 (van gogh, Drosophila) |
Vangl1 |
Kitenin, Lpp2, Stb2, Stbm |
-74.1kb | Anti-sense | Chr. 3 - 102155899 - 102205011 on NCBI Build GRCm38 |
Downstream | nerve growth factor | Ngf | Ngfb | 48.2kb | Sense | Chr. 3 - 102469919 - 102521013 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
