Assay Details
Gene Symbol: E430024P14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK051008.1 | - | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | T cell activation Rho GTPase activating protein | Tagap | 2610315E15Rik | Anti-sense | Chr. 17 - 7926000 - 7934897 on NCBI Build GRCm38 | |
Downstream | predicted gene 1604A | Gm1604a | 36.1kb | Sense | Chr. 17 - 7982518 - 8107845 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
