Assay Details
Gene Symbol: E430024P14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK051008.1 | - | 496 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | fibronectin type III domain containing 1 | Fndc1 |
1110027O12Rik, 1110051A18Rik, mKIAA1866 |
-55.2kb | Sense | Chr. 17 - 7738568 - 7827342 on NCBI Build GRCm38 |
Downstream | T cell activation Rho GTPase activating protein | Tagap | 2610315E15Rik | 39.0kb | Anti-sense | Chr. 17 - 7926000 - 7934897 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
