Assay Details
Gene Symbol: Erbb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003817.1 | NP_001003817.1 | 19 - 20 | 2484 | 70 | |
GenBank mRNA | AK031099.1 | - | 19 - 20 | 2352 | 70 | |
AK031542.1 | - | 11 - 12 | 1182 | 70 | ||
AK083669.1 | - | 18 - 19 | 2434 | 70 | ||
AK129487.1 | - | 19 - 20 | 2454 | 70 | ||
AK142296.1 | - | 19 - 20 | 2478 | 70 | ||
BC027080.1 | - | 1 - 2 | 96 | 70 | ||
BC046553.1 | - | 5 - 6 | 839 | 70 | ||
BC046811.1 | - | 18 - 19 | 2556 | 5702040 | 70 | |
BC053078.1 | - | 18 - 19 | 2556 | 5702040 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
