Assay Details
Gene Symbol: Ercc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007949.4 | NP_031975.2 | 12 - 13 | 1254 | 60 | |
GenBank mRNA | AK004652.1 | - | 8 - 9 | 906 | 60 | |
AK082323.1 | - | 5 - 6 | 507 | 60 | ||
AK082761.1 | - | 12 - 13 | 1255 | 60 | ||
AK141692.1 | - | 12 - 13 | 1273 | 60 | ||
AK145969.1 | - | 12 - 13 | 1263 | 60 | ||
BC034517.1 | - | 11 - 12 | 1190 | 4458839 | 60 | |
U97572.1 | - | 11 - 12 | 1238 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
