Assay Details
Gene Symbol: Dab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023118.5 | NP_075607.2 | 9 - 10 | 974 | 145 | |
GenBank mRNA | AK154716.1 | - | 9 - 10 | 958 | 145 | |
CD348957.1 | - | 7 - 8 | 497 | 145 | ||
U18869.1 | - | 9 - 10 | 907 | 145 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
