Assay Details
Gene Symbol: Vangl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177545.5 | NP_808213.2 | 2 - 3 | 359 | 141 | |
GenBank mRNA | AF481860.1 | - | 2 - 3 | 275 | 141 | |
AK135230.1 | - | 2 - 3 | 320 | 141 | ||
AK167743.1 | - | 2 - 3 | 241 | 141 | ||
BC138519.1 | - | 1 - 2 | 101 | 8861539 | 141 | |
BC138520.1 | - | 1 - 2 | 101 | 8861540 | 141 | |
BY729182.1 | - | 2 - 3 | 359 | 141 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF481860.1, AK135230.1, BC138519.1, BC138520.1, NM_177545.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
