Assay Details
Gene Symbol: Vangl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177545.5 | NP_808213.2 | 6 - 7 | 1377 | 71 | |
GenBank mRNA | AF481860.1 | - | 6 - 7 | 1293 | 71 | |
AK167743.1 | - | 7 - 8 | 1191 | 71 | ||
BC024687.1 | - | 1 - 2 | 36 | 71 | ||
BC138519.1 | - | 5 - 6 | 1119 | 8861539 | 71 | |
BC138520.1 | - | 5 - 6 | 1119 | 8861540 | 71 | |
BU610139.1 | - | 3 - 4 | 262 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
