Assay Details
Gene Symbol: Rdh12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030017.3 | NP_084293.1 | 5 - 6 | 785 | 67 | |
GenBank mRNA | AK020927.1 | - | 5 - 6 | 782 | 67 | |
AK039233.1 | - | 5 - 6 | 786 | 67 | ||
BC016204.1 | - | 5 - 6 | 732 | 4527851 | 67 | |
BC067213.1 | - | 4 - 5 | 1906 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
