Assay Details
Gene Symbol: Rgs13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153171.4 | NP_694811.1 | 1 - 2 | 104 | 139 | |
GenBank mRNA | AK043268.1 | - | 1 - 2 | 104 | 139 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK043268.1, NM_153171.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
