Assay Details
Gene Symbol: Rgs13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153171.4 | NP_694811.1 | 2 - 3 | 174 | 82 | |
GenBank mRNA | AF498319.1 | - | 2 - 3 | 223 | 82 | |
AK043268.1 | - | 2 - 3 | 174 | 82 | ||
BC049624.1 | - | 2 - 3 | 92 | 6585574 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF498319.1, AK043268.1, BC049624.1, NM_153171.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
