Assay Details
Gene Symbol: Pfkm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163487.1 | NP_001156959.1 | 14 - 15 | 1496 | 79 | |
NM_001163488.1 | NP_001156960.1 | 14 - 15 | 1432 | 79 | ||
NM_021514.4 | NP_067489.3 | 13 - 14 | 1449 | 79 | ||
GenBank mRNA | AF249894.1 | - | 13 - 14 | 79 | ||
AK002711.2 | - | 14 - 15 | 1399 | 79 | ||
AK049773.1 | - | 14 - 15 | 1404 | 79 | ||
BC005526.1 | - | 13 - 14 | 1413 | 3495173 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
