Assay Details
Gene Symbol: Nhp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011482.4 | NP_035612.2 | 2 - 3 | 234 | 82 | |
GenBank mRNA | AK004489.1 | - | 2 - 3 | 182 | 82 | |
AK134943.1 | - | 172 | 82 | |||
AK145391.1 | - | 2 - 3 | 236 | 82 | ||
AK145608.1 | - | 2 - 3 | 216 | 82 | ||
AK168808.1 | - | 2 - 3 | 204 | 82 | ||
AK168930.1 | - | 2 - 3 | 187 | 82 | ||
AK169103.1 | - | 2 - 3 | 218 | 82 | ||
BC026755.1 | - | 2 - 3 | 180 | 3154066 | 82 | |
BC054450.1 | - | 2 - 3 | 180 | 6491298 | 82 | |
BC083315.1 | - | 2 - 3 | 156 | 5714819 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
