Assay Details
Gene Symbol: Papss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289477.1 | NP_001276406.1 | 3 - 4 | 316 | 68 | |
NM_001289478.1 | NP_001276407.1 | 3 - 4 | 503 | 68 | ||
NM_011863.2 | NP_035993.1 | 2 - 3 | 326 | 68 | ||
GenBank mRNA | AK146507.1 | - | 2 - 3 | 199 | 68 | |
AK153296.1 | - | 2 - 3 | 196 | 68 | ||
AK168982.1 | - | 2 - 3 | 199 | 68 | ||
AK169117.1 | - | 3 - 4 | 378 | 68 | ||
AK169124.1 | - | 2 - 3 | 193 | 68 | ||
AK169204.1 | - | 3 - 4 | 378 | 68 | ||
BC066055.1 | - | 3 - 4 | 382 | 6833756 | 68 | |
U34883.1 | - | 2 - 3 | 203 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
