Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700028699.mm.1 |
TC0700004652.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700028699.mm.1 |
NM_001082961.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700028699.mm.1 |
NM_001082962.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700028699.mm.1 |
NM_013670.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700028699.mm.1 |
NM_033174.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700028709.mm.1 |
TC0700004652.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700028709.mm.1 |
NM_013670.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700028709.mm.1 |
NM_033174.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700028701.mm.1 |
TC0700004652.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700028701.mm.1 |
NM_001082961.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700028701.mm.1 |
NM_001082962.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700028701.mm.1 |
NM_013670.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700028701.mm.1 |
NM_033174.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700028698.mm.1 |
TC0700004652.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700028698.mm.1 |
NM_001082961.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700028698.mm.1 |
NM_001082962.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700028698.mm.1 |
NM_013670.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700028698.mm.1 |
NM_033174.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700028700.mm.1 |
TC0700004652.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700028700.mm.1 |
NM_001082961.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700028700.mm.1 |
NM_001082962.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700028700.mm.1 |
NM_013670.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700028700.mm.1 |
NM_033174.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700028713.mm.1 |
TC0700004652.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700028713.mm.1 |
NM_013670.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700028713.mm.1 |
NM_033174.4 |
Refseq |