Assay Details
Gene Symbol: Ints6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008715.2 | NP_032741.2 | 9 - 10 | 1627 | 127 | |
GenBank mRNA | AK046055.1 | - | 2 - 3 | 242 | 127 | |
BC059263.1 | - | 9 - 10 | 1627 | 6409580 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
