Assay Details
Gene Symbol: Cenpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134046.5 | NP_598807.2 | 2 - 3 | 666 | 85 | |
GenBank mRNA | AK013198.1 | - | 2 - 3 | 338 | 85 | |
AK018444.1 | - | 2 - 3 | 311 | 85 | ||
AK029226.1 | - | 2 - 3 | 667 | 85 | ||
AK134266.1 | - | 2 - 3 | 338 | 85 | ||
BC023148.1 | - | 2 - 3 | 309 | 4011571 | 85 | |
BC034399.1 | - | 2 - 3 | 325 | 4481047 | 85 | |
BC036278.1 | - | 2 - 3 | 309 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK013198.1, AK029226.1, AK134266.1, BC023148.1, BC034399.1, NM_134046.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
