Assay Details
Gene Symbol: Fermt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146054.2 | NP_666166.2 | 2 - 3 | 301 | 124 | |
GenBank mRNA | AK079588.1 | - | 2 - 3 | 234 | 124 | |
AK145030.1 | - | 2 - 3 | 243 | 124 | ||
AK165608.1 | - | 2 - 3 | 272 | 124 | ||
AK166552.1 | - | 2 - 3 | 240 | 124 | ||
BC033436.1 | - | 2 - 3 | 222 | 4950810 | 124 | |
BQ179963.1 | - | 1 - 2 | 101 | 124 | ||
BY301704.1 | - | 2 - 3 | 301 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
