Assay Details
Gene Symbol: Akt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011785.3 | NP_035915.3 | 6 - 7 | 675 | 87 | |
GenBank mRNA | AF124142.1 | - | 6 - 7 | 665 | 87 | |
BC066861.1 | - | 6 - 7 | 675 | 30089997 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
