Assay Details
Gene Symbol: Rps6kc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178775.4 | NP_848890.3 | 11 - 12 | 2898 | 78 | |
GenBank mRNA | AK044809.1 | - | 8 - 9 | 2617 | 78 | |
BC058403.1 | - | 2 - 3 | 1597 | 6812243 | 78 | |
BG079911.2 | - | 1 - 2 | 272 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
