Assay Details
Gene Symbol: Pde10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290707.1 | NP_001277636.1 | 10 - 11 | 1181 | 123 | |
NM_011866.2 | NP_035996.2 | 9 - 10 | 1135 | 123 | ||
GenBank mRNA | AF110507.1 | - | 9 - 10 | 1006 | 123 | |
AK039249.1 | - | 10 - 11 | 1181 | 123 | ||
AK147690.1 | - | 8 - 9 | 1052 | 123 | ||
AK162221.1 | - | 9 - 10 | 790 | 123 | ||
AK162804.1 | - | 7 - 8 | 676 | 123 | ||
AY360383.1 | - | 9 - 10 | 1135 | 123 | ||
BC113201.1 | - | 9 - 10 | 1001 | 40048238 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
