Assay Details
Gene Symbol: Trpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039104.2 | NP_001034193.2 | 5 - 6 | 878 | 72 | |
NM_018752.3 | NP_061222.3 | 5 - 6 | 771 | 72 | ||
GenBank mRNA | AF047714.1 | - | 5 - 6 | 640 | 72 | |
AK044507.1 | - | 5 - 6 | 869 | 72 | ||
AK148094.1 | - | 5 - 6 | 676 | 72 | ||
AK148193.1 | - | 5 - 6 | 679 | 72 | ||
AK148268.1 | - | 5 - 6 | 625 | 72 | ||
AK148275.1 | - | 5 - 6 | 681 | 72 | ||
AK149290.1 | - | 5 - 6 | 800 | 72 | ||
AK164395.1 | - | 5 - 6 | 681 | 72 | ||
AK164423.1 | - | 5 - 6 | 688 | 72 | ||
AK190255.1 | - | 1 - 2 | 60 | 72 | ||
AK190666.1 | - | 1 - 2 | 60 | 72 | ||
AK190766.1 | - | 1 - 2 | 60 | 72 | ||
AK191673.1 | - | 60 | 72 | |||
AK191720.1 | - | 1 - 2 | 60 | 72 | ||
AK192224.1 | - | 1 - 2 | 61 | 72 | ||
AK193404.1 | - | 1 - 2 | 60 | 72 | ||
AK193726.1 | - | 1 - 2 | 60 | 72 | ||
AK194458.1 | - | 1 - 2 | 60 | 72 | ||
AK194882.1 | - | 1 - 2 | 60 | 72 | ||
AK195005.1 | - | 1 - 2 | 128 | 72 | ||
AK195099.1 | - | 1 - 2 | 60 | 72 | ||
AK197512.1 | - | 1 - 2 | 60 | 72 | ||
AK197999.1 | - | 1 - 2 | 60 | 72 | ||
AK198239.1 | - | 1 - 2 | 60 | 72 | ||
AK199238.1 | - | 1 - 2 | 60 | 72 | ||
AK199411.1 | - | 1 - 2 | 60 | 72 | ||
AK199444.1 | - | 1 - 2 | 60 | 72 | ||
AK200256.1 | - | 1 - 2 | 60 | 72 | ||
AK201500.1 | - | 1 - 2 | 60 | 72 | ||
AK201639.1 | - | 2 - 3 | 134 | 72 | ||
AK201763.1 | - | 1 - 2 | 60 | 72 | ||
AK201774.1 | - | 1 - 2 | 60 | 72 | ||
AK202394.1 | - | 1 - 2 | 60 | 72 | ||
AK202694.1 | - | 1 - 2 | 60 | 72 | ||
AK203152.1 | - | 1 - 2 | 60 | 72 | ||
AK203404.1 | - | 1 - 2 | 60 | 72 | ||
AK204671.1 | - | 1 - 2 | 60 | 72 | ||
AK205104.1 | - | 1 - 2 | 60 | 72 | ||
AK205189.1 | - | 1 - 2 | 60 | 72 | ||
AK210565.1 | - | 1 - 2 | 60 | 72 | ||
AK212948.1 | - | 1 - 2 | 60 | 72 | ||
AK216555.1 | - | 1 - 2 | 60 | 72 | ||
AK219405.1 | - | 1 - 2 | 60 | 72 | ||
AY180104.2 | - | 5 - 6 | 878 | 72 | ||
BC082560.1 | - | 5 - 6 | 647 | 72 | ||
BC085168.1 | - | 4 - 5 | 450 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
