Assay Details
Gene Symbol: Arnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007488.3 | NP_031514.3 | 3 - 4 | 347 | 63 | |
GenBank mRNA | AB093225.1 | - | 5 - 6 | 527 | 63 | |
AK032628.1 | - | 3 - 4 | 328 | 63 | ||
BC054546.1 | - | 3 - 4 | 268 | 6409636 | 63 | |
D63644.1 | - | 3 - 4 | 223 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
