Assay Details
Gene Symbol: Tbck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK034533.1 | - | 10 - 11 | 1160 | 74 | |
AK052841.1 | - | 10 - 11 | 1172 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
