Assay Details
Gene Symbol: Xpo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020506.1 | NP_065252.1 | 14 - 15 | 1972 | 78 | |
GenBank mRNA | AF145021.1 | - | 14 - 15 | 1972 | 78 | |
AK084907.1 | - | 15 - 16 | 2157 | 78 | ||
AK129430.1 | - | 14 - 15 | 1967 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
