Assay Details
Gene Symbol: Il3ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008369.1 | NP_032395.1 | 10 - 11 | 1200 | 102 | |
GenBank mRNA | BC049889.1 | - | 9 - 10 | 1600 | 102 | |
FJ550346.1 | - | 7 - 8 | 782 | 102 | ||
X64534.1 | - | 10 - 11 | 1200 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
