Assay Details
Gene Symbol: Il3ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008369.1 | NP_032395.1 | 3 - 4 | 355 | 70 | |
GenBank mRNA | BC049889.1 | - | 2 - 3 | 755 | 70 | |
BC077736.1 | - | 2 - 3 | 296 | 70 | ||
X64534.1 | - | 3 - 4 | 355 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
